Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532084
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532084 Supplier Invitrogen™ Supplier No. PA532084
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HeLa. Predicted reactivity: Mouse (83%), Rat (83%), Chimpanzee (98%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Armadillo/beta-catenin (CTNNB1; MIM 116806)-like (ARM) domains are imperfect 45-amino acid repeats involved in protein-protein interactions. ARM domain-containing proteins, such as ARMC3, function in signal transduction, development, cell adhesion and mobility, and tumor initiation and metastasis (Li et al., 2006 [PubMed 16915934]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMC3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ARMC3
Gene Accession No. Q5W041
Gene Alias 4921513G22Rik; armadillo repeat containing 3; armadillo repeat-containing protein 3; Armc3; Beta-catenin-like protein; cancer/testis antigen 81; CT81; KU-CT-1
Gene Symbols ARMC3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ARMC3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219681
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.