Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMC6 Monoclonal Antibody (2H8)
GREENER_CHOICE

Catalog No. PIMA522880
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522880 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522880 Supplier Invitrogen™ Supplier No. MA522880
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MVSKRIAQET FDAAVRENIE EFAMGPEEAV KEAVEQFESQ GVDLSNIVKT APKVSADGSQ EPTHDILQML SDLQESVASS RPQEVSAYLT RFCDQCKQDK ACRFLAAQKG AYPIIFTAWK LATAGDQGLL LQSLNALSVL TDGQPDLLDA QGLQLLVATL TQNADEADLT CSGIRCVRHA CLKHEQNRQD LVKAGVLPLL TGAITHHGHH TDVVREACWA LRVMTFDDDI RVPFGHAHNH AKMIVQENKG LKVLIEATKA FLDNPGILSE LCGTLSRLAI RNEFCQEVVD LGGLSILVSL LADCNDHQMR DQSGVQELVK QVLSTLRAIA GNDDVKDAIV RAGGTESIVA AMTQHLTSPQ VCEQSCAALC FLALRKPDNS RIIVEGGGAV AALQAMKAHP QKAGVQKQAC MLIRNLVAHG QAFSKPILDL GAEALIMQAR SAHRDCEDVA KAALRDLGCH VELRELWTGQ RGNLAP.

The function of this gene's protein product has not been determined. A related protein in mouse suggests that this protein has a conserved function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMC6
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2H8
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Armc6
Gene Accession No. Q6NXE6
Gene Alias 2410153K17Rik; armadillo repeat containing 6; armadillo repeat-containing protein 6; Armc6; AW554412; R30923_1; RGD1306691
Gene Symbols Armc6
Host Species Mouse
Immunogen ARMC6 (AAH67125, 1 a.a. ~ 476 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93436
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.