Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106605 Supplier Invitrogen™ Supplier No. PA5106605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARMC6.

The function of this gene's protein product has not been determined. A related protein in mouse suggests that this protein has a conserved function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMC6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Armc6
Gene Accession No. Q6NXE6, Q8BNU0
Gene Alias 2410153K17Rik; armadillo repeat containing 6; armadillo repeat-containing protein 6; Armc6; AW554412; R30923_1; RGD1306691
Gene Symbols Armc6
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARMC6(Accession Q6NXE6), corresponding to amino acid residues H444-G494.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306352, 76813, 93436
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.