Learn More
ARMC6 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305645AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
The function of this gene′s protein product has not been determined. A related protein in mouse suggests that this protein has a conserved function.Specifications
ARMC6 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q6NXE6 | |
ARMC6 | |
Between 451 and 501 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation | |
Unconjugated | |
ARMC6 | |
Armadillo repeat-containing protein 6, ARMC6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
93436 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.