Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMC7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555021
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555021 Supplier Invitrogen™ Supplier No. PA555021
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAQKPKVDPH VGRLGYLQAL VTEFQETQSQ DAKEQVLANL ANFAYDPSNY EYLRQLQVLD LFLDSLSEEN ETLVEFAIGG LCNLCPDRAN KEHILHAGG Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.

ARMC7 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMC7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARMC7
Gene Accession No. Q3UJZ3, Q9H6L4
Gene Alias A630084C13; A630084C13rik; armadillo repeat containing 7; armadillo repeat-containing protein 7; Armc7; BC023126; RGD1310043
Gene Symbols ARMC7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARMC7. Recombinant protein control fragment (Product #RP-93251).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 276905, 287827, 79637
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.