Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARMC7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565763
Description
Immunogen sequence: LIINCLSSPNE ETVLSAITTL MHLSPPGRSF LPELTATPVV QCMLRFSLSA SARLRNLAQI FLEDFCSPRQ VAEARSRQAH SALGI Highest antigen sequence identity to the following orthologs - mouse 81%, rat 80%.
ARMC7 is a protein coding gene.
Specifications
ARMC7 | |
Polyclonal | |
Unconjugated | |
ARMC7 | |
A630084C13; A630084C13rik; armadillo repeat containing 7; armadillo repeat-containing protein 7; Armc7; BC023126; RGD1310043 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79637 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H6L4 | |
ARMC7 | |
Recombinant Human ARMC7. Recombinant protein control fragment (Product #RP-106442). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction