Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMC8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565394 Supplier Invitrogen™ Supplier No. PA565394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEGEHNIEVKE QTLCILANIA DGTTAKDLIM TNDDILQKIK YYMGHSHVKL QLAAMFCISN LIWNEEEGSQ ERQDKLRDMG IVDILHKLSQ SPDSNL Highest antigen sequence identity to the following orthologs - mouse 97%, rat 96%.

ARMC8 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMC8
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARMC8
Gene Accession No. Q8IUR7
Gene Alias 1200015K23Rik; armadillo repeat containing 8; armadillo repeat-containing protein 8; Armc8; GID complex subunit 5, VID28 homolog; GID5; HSPC056; RGD1310477; S863-2; VID28
Gene Symbols ARMC8
Host Species Rabbit
Immunogen Recombinant Human ARMC8. Recombinant protein control fragment (Product #RP-106077).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25852
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.