Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARMC9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555370
Description
Immunogen sequence: VALLMNLCLR STGKNMCAKV AGLVLKVLSD LLGHENHEIQ PYVNGALYSI LSVPSIREEA RAMGMEDILR CFIKEGNAEM IRQIEFIIKQ LNSEELPDGV LESD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Acts as a positive regulator of hedgehog (Hh)signaling. Involved in ciliogenesis. May participate in the trafficking and/or retention of GLI2 and GLI3 proteins at the ciliary tip. [UniProt].
Specifications
| ARMC9 | |
| Polyclonal | |
| Unconjugated | |
| ARMC9 | |
| ARM; armadillo repeat containing 9; Armadillo repeat-containing protein 9; armadillo/beta-catenin-like repeats; ARMC9; KIAA1868; KU-MEL-1; lisH domain-containing protein ARMC9; melanoma/melanocyte specific protein KU-MEL-1; melanoma/melanocyte-specific tumor antigen KU-MEL-1; NS21 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80210 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z3E5 | |
| ARMC9 | |
| Recombinant protein corresponding to Human ARMC9. Recombinant protein control fragment (Product #RP-92523). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction