Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMC9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598569
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598569 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598569 Supplier Invitrogen™ Supplier No. PA598569
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARMC9 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Acts as a positive regulator of hedgehog (Hh)signaling. Involved in ciliogenesis. May participate in the trafficking and/or retention of GLI2 and GLI3 proteins at the ciliary tip. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMC9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ARMC9
Gene Accession No. Q7Z3E5
Gene Alias ARM; armadillo repeat containing 9; Armadillo repeat-containing protein 9; armadillo/beta-catenin-like repeats; ARMC9; KIAA1868; KU-MEL-1; lisH domain-containing protein ARMC9; melanoma/melanocyte specific protein KU-MEL-1; melanoma/melanocyte-specific tumor antigen KU-MEL-1; NS21
Gene Symbols ARMC9
Host Species Rabbit
Immunogen Recombinant Human LisH domain-containing protein ARMC9 protein (401-665AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80210
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.