Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMCX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565687 Supplier Invitrogen™ Supplier No. PA565687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TKFAHQSEASF PVEDESRKQT RTGEKTRPWS CRCKHEANMD PRDLEKLICM IEMTEDPSVH EIANNALYNS ADYSYSHEVV RNVGGI Highest antigen sequence identity to the following orthologs - mouse 64%, rat 63%.

The product of this gene belongs to the armadillo repeat-containing family of proteins, which interact with other proteins in a variety of cellular processes. The function of this family member is currently unknown. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq, Jan 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMCX4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARMCX4
Gene Accession No. Q5H9R4
Gene Alias armadillo repeat containing, X-linked 4; armadillo repeat containing, X-linked 4 pseudogene; armadillo repeat-containing X-linked protein 4; ARMCX4; CXorf35; GASP4
Gene Symbols ARMCX4
Host Species Rabbit
Immunogen Recombinant Human ARMCX4. Recombinant protein control fragment (Product #RP-106361).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131755
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.