Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARMCX4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549083
Description
The product of this gene belongs to the armadillo repeat-containing family of proteins, which interact with other proteins in a variety of cellular processes. The function of this family member is currently unknown. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq, Jan 2012]
Specifications
ARMCX4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q5H9R4 | |
ARMCX4 | |
KLH conjugated synthetic peptide between 114-140 amino acids from the Central region of human ARMCX4. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ARMCX4 | |
Armadillo repeat-containing X-linked protein 4, ARMCX4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100131755 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction