Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARMCX6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542600
Description
Peptide sequence: TMARPWTEDG DWTEPGAPGG TEDRPSGGGK ANRAHPIKQR PFPYEHKNTW Sequence homology: Dog: 79%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 100%; Rat: 93%.
May regulate the dynamics and distribution of mitochondria in neural cells. [UniProt].
Specifications
ARMCX6 | |
Polyclonal | |
Unconjugated | |
ARMCX6 | |
arm protein lost in epithelial cancers, X chromosome, 3; armadillo repeat containing X-linked 6; armadillo repeat containing, X-linked 6; Armcx6; AW060994; GASP10; protein ARMCX6 | |
Rabbit | |
Protein A | |
RUO | |
54470 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7L4S7 | |
ARMCX6 | |
Synthetic peptide directed towards the N-terminal of human ARMCX6 (aa 64-113). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction