Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Armenian Hamster anti-Integrin alpha 2/CD49b, Clone: HMa2, FITC, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
$585.00
Specifications
Antigen | Integrin alpha 2/CD49b |
---|---|
Clone | HMa2 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14400201
![]() |
Novus Biologicals
NBP1440020.1MG |
0.1 mg |
Each of 1 for $585.00
|
|
|||||
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Mouse samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 2/CD49b | |
0.2 mg/mL | |
Flow Cytometry | |
FITC | |
RUO | |
Mouse | |
alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2 | |
ITGA2 | |
IgG | |
Affinity Purified |
HMa2 | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Armenian Hamster | |
Cellular Markers, Innate Immunity, Signal Transduction | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
3673 | |
The immunogen for this antibody was CD49b. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title