Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 2/CD49b Armenian Hamster anti-Mouse, Clone: HMa2, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
$182.00 - $611.00
Specifications
| Antigen | Integrin alpha 2/CD49b |
|---|---|
| Clone | HMa2 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunoprecipitation, Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018544
![]() |
Novus Biologicals
NBP1434020.025MG |
0.025 mg |
Each for $182.00
|
|
|||||
NBP14340205
![]() |
Novus Biologicals
NBP1434020.05MG |
0.05 mg |
Each for $305.00
|
|
|||||
NBP14340201
![]() |
Novus Biologicals
NBP1434020.1MG |
0.1 mg |
Each for $464.00
|
|
|||||
NBP1434025
![]() |
Novus Biologicals
NBP1434020.5MG |
0.5 mg |
Each for $611.00
|
|
|||||
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| Integrin alpha 2/CD49b | |
| 0.5 mg/mL | |
| Immunoprecipitation, Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Mouse | |
| alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2 | |
| ITGA2 | |
| IgG | |
| Affinity Purified |
| HMa2 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Armenian Hamster | |
| Cellular Markers, Innate Immunity, Signal Transduction | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 3673 | |
| The immunogen for this antibody was CD49b. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title