Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Armenian Hamster IgG, Clone: 299Arm, PE-Cy5, Isotype control, Novus Biologicals™
SDP

Catalog No. NBP14307101 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14307101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP14307101 Supplier Novus Biologicals Supplier No. NBP1430710.1MG

Armenian Hamster Monoclonal Antibody

IgG Monoclonal specifically detects IgG in Armenian Hamster samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG
Applications Flow Cytometry
Classification Monoclonal
Clone 299Arm
Conjugate PE-Cyanine5
Dilution Flow Cytometry 1:10 - 1:1000
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Gene Alias immunoglobulin heavy constant gamma 1 (G1m marker)
Host Species Armenian Hamster
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This 299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood. This should be used at the same concentration as the experimental antibody.
Target Species Armenian Hamster
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.