Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Armenian Hamster IgG Isotype Control (299Arm), Novus Biologicals™
SDP

Catalog No. NBP259616 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259616 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259616 Supplier Novus Biologicals Supplier No. NBP259616
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

IgG Monoclonal specifically detects IgG in Armenian Hamster samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, In vitro assay, Control.

Specifications

Antigen IgG
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunocytochemistry
Classification Monoclonal
Clone 299Arm
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Control, Immunofluorescence 1:10-1:500, Immunocytochemistry 1:10-1:500
Formulation PBS, pH 7.2 with 0.09% Sodium Azide
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Armenian Hamster
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Gene ID (Entrez) 3500
Target Species Armenian Hamster
Content And Storage Store at 4°C.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.