Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMER Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520061
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520061 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520061 Supplier Invitrogen™ Supplier No. PA520061
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is mouse small intestine tissue lysate. PA5-20061 can be used with blocking peptide PEP-0180.

Apoptosis is important for normal development and tissue homeostasis. It is mediated by various caspases and ultimately results in the activation of endogenous endonucleases that degrade cellular DNA. Although apoptosis induced by endoplasmic reticulum (ER) stress is thought to be mediated by caspase-12, other caspases such as caspase-9 are also thought to be activated following ER stress. Recently, ARMER, a novel integral ER-membrane protein was shown to protect cells from ER stress-induced apoptosis. Analysis of the caspase proteolytic cascade suggests that ARMER acts by inhibiting caspase-9 activity, although the mechanism for this remains unknown. It should be noted that ARMER is not related to the inhibitor of apoptosis proteins (IAP) family and does not contain any baculoviral IAP repeat (BIR) domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMER
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ARL6IP1
Gene Accession No. Q15041, Q9JKW0
Gene Alias ADP ribosylation factor like GTPase 6 interacting protein 1; ADP-ribosylation factor GTPase 6 interacting protein 1; ADP-ribosylation factor like GTPase 6 interacting protein 1; ADP-ribosylation factor-like 6 interacting protein; ADP-ribosylation factor-like 6 interacting protein 1; ADP-ribosylation factor-like protein 6-interacting protein 1; ADP-ribosylation factor-like protein 6-interacting protein 1-like protein; ADP-ribosylation-like factor 6 interacting protein; ADP-ribosylation-like factor 6-interacting protein; AIP1; aip-1; AIP-6; AL022945; apoptotic regulator in the membrane of the endoplasmic reticulum; ARL-6-interacting protein 1; Arl6ip; arl6ip protein; Arl6ip1; ARMER; AU042858; C85138; KIAA0069; mKIAA0069; Protein TBX2; SPG61; zgc:56593; zgc:77873
Gene Symbols ARL6IP1
Host Species Rabbit
Immunogen A peptide corresponding to 15 amino acids near the C-terminus of human ARMER.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23204, 54208
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.