Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMER Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560977 Supplier Invitrogen™ Supplier No. PA560977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FGSNKWTTEQ QQRFHEICSN LVKTRRRAVG WWKRLFTLKE E Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 100%.

Apoptosis is important for normal development and tissue homeostasis. It is mediated by various caspases and ultimately results in the activation of endogenous endonucleases that degrade cellular DNA. Although apoptosis induced by endoplasmic reticulum (ER) stress is thought to be mediated by caspase-12, other caspases such as caspase-9 are also thought to be activated following ER stress. Recently, ARMER, a novel integral ER-membrane protein was shown to protect cells from ER stress-induced apoptosis. Analysis of the caspase proteolytic cascade suggests that ARMER acts by inhibiting caspase-9 activity, although the mechanism for this remains unknown. It should be noted that ARMER is not related to the inhibitor of apoptosis proteins (IAP) family and does not contain any baculoviral IAP repeat (BIR) domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMER
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARL6IP1
Gene Accession No. Q15041
Gene Alias ADP ribosylation factor like GTPase 6 interacting protein 1; ADP-ribosylation factor GTPase 6 interacting protein 1; ADP-ribosylation factor like GTPase 6 interacting protein 1; ADP-ribosylation factor-like 6 interacting protein; ADP-ribosylation factor-like 6 interacting protein 1; ADP-ribosylation factor-like protein 6-interacting protein 1; ADP-ribosylation factor-like protein 6-interacting protein 1-like protein; ADP-ribosylation-like factor 6 interacting protein; ADP-ribosylation-like factor 6-interacting protein; AIP1; aip-1; AIP-6; AL022945; apoptotic regulator in the membrane of the endoplasmic reticulum; ARL-6-interacting protein 1; Arl6ip; arl6ip protein; Arl6ip1; ARMER; AU042858; C85138; KIAA0069; mKIAA0069; Protein TBX2; SPG61; zgc:56593; zgc:77873
Gene Symbols ARL6IP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARMER. Recombinant protein control fragment (Product #RP-91178).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23204
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.