Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARMET Rabbit anti-Human, Mouse, Rat, FITC, Polyclonal, FabGennix

Catalog No. 502207662
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7662 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7662 Supplier Fabgennix Inc Supplier No. MANFFITC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARMET Polyclonal antibody specifically detects ARMET in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen ARMET
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate FITC
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene MANF
Gene Accession No. P0C5H9, P55145, Q9CXI5
Gene Alias 3230402M22Rik; AA407711; AA408789; AA673178; Arginine-rich protein; arginine-rich, mutated in early stage tumors; ARMET; ARP; D17914; D18Mgi17; Manf; mesencephalic astrocyte derived neurotrophic factor; mesencephalic astrocyte-derived neurotrophic factor; MGC142148; MGC142150; Protein ARMET
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 80-120 on human Mesencephalic astrocyte-derived neurotrophic factor.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315989, 74840, 7873
Target Species Human, Mouse, Rat
Content And Storage -20° C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.