Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARMH4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP170435
SDP
View Documents
Novus Biologicals
NBP170435
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

ARMH4 Polyclonal specifically detects ARMH4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Western Blot
Unconjugated
RUO
145407
Synthetic peptides corresponding to C14ORF37 The peptide sequence was selected from the N terminal of C14ORF37. Peptide sequence EIAHVHAEKGQSDKMNTDDLENSSVTSKQTPQLVVSEDPMMMSAVPSATS.
Primary
Polyclonal
Rabbit
c14_5376, chromosome 14 open reading frame 37
C14ORF37
IgG
84 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.