Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ArmorGrip™ Rough Texture Nitrile Coated Gloves - Safety Cuff

Catalog No. 19161897 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-161-897 Medium
19-816-808 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-161-897 Supplier PIP™ Supplier No. 563145/M
This item is not returnable. View return policy

Armor™ Series gloves combine low-coating deposit weight and high abrasion resistance to provide the wear of heavier, bulkier nitrile coated gloves without sacrificing flexibility and tactile sensitivity.

  • Protection from physical hazards such as cuts, punctures, snags and abrasions
  • Terry cloth palm coated with rough textured nitrile enhances grip capabilities
  • Safety cuff helps provides additional wrist protection, and allows for fast removal
  • ActiFresh™ treatment reduces germs and odors
  • Manufactured in ISO 9001:2000/ISO 14001:2000 facilities

Specifications

Coated Area Palm, Fingers, Knuckles
Coating Material Nitrile
Color Blue
Exterior Finish Fully Textured
Hand Specific
Product Type Dip-coated Gloves
Size Medium
For Use With (Application) General Assembly, Fabrication, Parts Handling, and Maintenance
Glove Cuff Color Natural
Glove Cuff Style Safety
Liner Material Terry Cloth
Series 56-3145
Length (English) 10.8 in.
Length (Metric) 27.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.