Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587689
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587689 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587689 Supplier Invitrogen™ Supplier No. PA587689
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLRLYPGPMV TEAEGKGGPE MASLSSSVVP VSFISTLRES VLDPGVGGEG ASDKQRSKLS LSHSMIPAAK IHTELCLPAF FSPAGTQRRF QQPQHHLTLS IIHTAAR; Positive Samples: Mouse eye; Cellular Location: Cytoplasm.

This gene encodes a small secreted protein specific to primates. This protein is a component of the choroidal extracellular matrix of the eye. Mutations in this gene are associated with age-related macular degeneration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Alias age-related maculopathy susceptibility 2; age-related maculopathy susceptibility protein 2; ARMD8; ARMS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-107 of human ARMS2 (NP_0010931371).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.