Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566904
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566904 Supplier Invitrogen™ Supplier No. PA566904
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSSSVVPVSFI STLRESVLDP GVGGEGASD Highest antigen sequence identity to the following orthologs - mouse 42%, rat 41%.

This gene encodes a small secreted protein specific to primates. This protein is a component of the choroidal extracellular matrix of the eye. Mutations in this gene are associated with age-related macular degeneration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMS2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARMS2
Gene Accession No. P0C7Q2
Gene Alias age-related maculopathy susceptibility 2; age-related maculopathy susceptibility protein 2; ARMD8; ARMS2
Gene Symbols ARMS2
Host Species Rabbit
Immunogen Recombinant Human ARMS2. Recombinant protein control fragment (Product #RP-107556).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387715
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.