Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARMT1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP261238
Description
CHMP7 Polyclonal specifically detects CHMP7 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ARMT1 | |
Polyclonal | |
Western Blot 1:1000, Immunoprecipitation 20 ul/mg lysate | |
ARMT1 | |
between 391 and 441 | |
0.1 mL | |
79624 | |
Store the unopened product at 2 - 8degrees C. Protect from light. Do not use past expiration date. |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
Rabbit | |
Affinity Purified | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction