Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARMX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552183
Description
Immunogen sequence: DEESTDTSEI GVETVKGAKT NAGAGSGAKL QGDSEVKPEV SLGLEDCPGV KEKAHSGSHS GGGLEAKAKA LFNTLKEQAS AKAGKGARVG TISGNRTLAP SLPCP Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 57%.
This gene encodes a member of the ALEX family of proteins and may play a role in tumor suppression. The encoded protein contains a potential N-terminal transmembrane domain and two Armadillo (arm) repeats. Other proteins containing the arm repeat are involved in development, maintenance of tissue integrity, and tumorigenesis. This gene is closely localized with other family members, including ALEX2 and ALEX3, on the X chromosome.
Specifications
ARMX1 | |
Polyclonal | |
Unconjugated | |
ARMCX1 | |
3010033I09Rik; AD032; ALEX1; ARM protein lost in epithelial cancers on chromosome X 1; arm protein lost in epithelial cancers, X chromosome, 1; armadillo repeat containing, X-linked 1; armadillo repeat-containing X-linked protein 1; ARMCX1; GASP7; Protein ALEX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51309 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P291 | |
ARMCX1 | |
Recombinant protein corresponding to Human ARMX1. Recombinant protein control fragment (Product #RP-91893). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction