Learn More
Invitrogen™ ARMX2 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ARMCX2 protein.
Specifications
Specifications
| Antigen | ARMX2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ARMCX2 |
| Gene Accession No. | Q6A058, Q7L311 |
| Gene Alias | 3230401N03Rik; AI043003; ALEX2; ALEX2 protein; ARM protein lost in epithelial cancers on chromosome X 2; armadillo repeat containing, X-linked 2; armadillo repeat protein ALEX2; armadillo repeat-containing X-linked protein 2; ARMCX2; GASP9; Kiaa0512; Protein ALEX2 |
| Gene Symbols | ARMCX2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.