Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552201
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552201 Supplier Invitrogen™ Supplier No. PA552201
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RDQTKKRMAK PKNRAVAGTG ARARAGLRAG FTIDLGSGFS PPTPVRAEAE DRAQDEASAL DTVGAEAVAP AASSAEAQSG AGSQAQEADG AGVGPKAESV VGAAMASAIA PPPGVTEALG AAEAPAMAGA PKVAEAPREA ETSRA Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 55%.

The armadillo (ARM) repeat family of proteins are related to the Drosophila melanogaster armadillo protein, a protein essential for wingless signal transduction. ARM proteins are involved in a variety of processes such as cell migration, cell proliferation, tissue maintenance and tumorigenesis, and they also function in signal transduction and the maintenance of overall cell structure. ARMCX2 (armadillo repeat containing, X-linked 2), also known as ALEX2, is a 632 amino acid single-pass membrane protein that contains three ARM repeats and is highly expressed in testis, brain, ovary, heart, colon, spleen and prostate, where it is thought to play a role in tumor suppression. The gene encoding ARMCX2 maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1, 000 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMX2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARMCX2
Gene Accession No. Q7L311
Gene Alias 3230401N03Rik; AI043003; ALEX2; ALEX2 protein; ARM protein lost in epithelial cancers on chromosome X 2; armadillo repeat containing, X-linked 2; armadillo repeat protein ALEX2; armadillo repeat-containing X-linked protein 2; ARMCX2; GASP9; Kiaa0512; Protein ALEX2
Gene Symbols ARMCX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARMX2. Recombinant protein control fragment (Product #RP-92150).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9823
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.