Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARMX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551376 Supplier Invitrogen™ Supplier No. PA551376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAEGGSGDVD DAGDCSGARY NDWSDDDDDS NESKSIVWYP PWARIGTEAG TRARARARAR ATRARRAVQK RASPNSDDTV LSPQELQKVL CLVEMSEKPY ILEAALIALG NNAAY Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 98%.

This gene encodes a member of the ALEX family of proteins which may play a role in tumor suppression. The encoded protein contains a potential N-terminal transmembrane domain and a single Armadillo (arm) repeat. Other proteins containing the arm repeat are involved in development, maintenance of tissue integrity, and tumorigenesis. This gene is closely localized with other family members on the X chromosome. Three transcript variants encoding the same protein have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARMX3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARMCX3
Gene Accession No. Q9UH62
Gene Alias 1200004E24Rik; AI450003; ALEX3; ARM protein lost in epithelial cancers on chromosome X 3; arm protein lost in epithelial cancers, X chromosome, 3; armadillo repeat containing X-linked 3; armadillo repeat containing, X-linked 3; armadillo repeat-containing X-linked protein 3; armadillo repeat-containing X-linked protein 3-like protein; Armcx3; BM-017; dJ545K15.2; DKFZp781N1954; GASP6; KIAA0443; MGC12199; Protein ALEX3; UNQ2517/PRO6007
Gene Symbols ARMCX3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARMX3. Recombinant protein control fragment (Product #RP-91936).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51566
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.