Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ARN 11391
GREENER_CHOICESDP

Catalog No. 790950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-095-0 50 mg
79-091-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 79-095-0 Supplier Tocris Bioscience™ Supplier No. 7909/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potentiator of the IP3-gated calcium channel ITPR1

ARN 11391 is a potentiator of the IP3-gated calcium channel ITPR1 (Inositol 1,4,5-trisphosphate (IP3) receptor type 1). It amplifies intracellular calcium ion signaling by acting directly on ITPR1. ARN 11391 also potentiates calcium signaling in cells expressing mutant ITPR1 which is associated with SCA29, a form of spinocerebellar ataxia.

Specifications

Chemical Name or Material 2-Ethyl-4,5,6,7-tetrahydro-N-[2-[(tetrahydro-2 H-pyran-2-yl)methoxy]phenyl]-2 H-indazole-3-carboxamide
CAS 1214569-31-9
Quantity 50 mg
Target IP3R1
Molecular Formula C22H29N3O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.