Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ARN 14494
GREENER_CHOICE

Catalog No. 713410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-341-0 10 mg
71-345-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 71-341-0 Supplier Tocris Bioscience™ Supplier No. 7134/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent serine palmitoyltransferase inhibitor

ARN 14494 is a potent serine palmitoyltransferase inhibitor (IC50 = 27.3 nM). ARN 14494 inhibits synthesis of long chain ceramides and dihydroceramides in an in vitro model of Alzheimer's diease. The compound prevents the synthesis of proinflammatory cytokines and the oxidative stress-related enzymes iNOS and COX-2 in mouse primary cortical astrocytes. ARN 14494 is neuroprotective against β-amyloid 1-42 induced neurotoxicity in primary cortical neurons co-cultured with astrocytes.

Specifications

Chemical Name or Material rel-1-[1-[2-[(3 aR,6 AS)-Hexahydrocyclopenta[c]pyrrol-2(1 H)-yl]-2-oxoethyl]-4-piperidinyl]-2,3-dihydro-N-methyl-2-oxo-1 H-indole-5-carboxamide
CAS 1037834-43-7
Quantity 10 mg
Target Other Transferase Inhibitors
Molecular Formula C24H32N4O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.