Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ARN 3236
GREENER_CHOICESDP

Catalog No. 751150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-115-0 50 mg
75-111-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 75-115-0 Supplier Tocris Bioscience™ Supplier No. 7511/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent inhibitor of salt-inducible kinases

ARN 3236 is a potent of salt-inducible kinase 2 (SIK2) inhibitor (IC50 values are <1 nM, 6.63 nM and 21.63 nM for SIK2, SIK3 and SIK1 respectively). ARN 3236 inhibits centrosome separation by uncoupling the centrosome from the nucleus. It also induces cell-cycle arrest and apoptosis by attenuating the AKT/survivin pathway. ARN 3236 inhibits growth and enhances Taxol (Cat. No. 1097) sensitivity in ovarian cancer cell lines in vitro and in vivo. In human myeloid cells, ARN 3236 blocks TNF and induces IL-10 upon LPS stimulation; it also reduces the production of IL-1β upon activation of TLR4 and TLR2 signaling. In depression models, ARN 3236 exhibits antidepressant-like effects via the hippocampal CRTC1-CREB-BDNF pathway. ARN 3236 penetrates the blood-brain barrier, and it is orally bioavailable.

Specifications

Chemical Name or Material 3-(2,4-Dimethoxyphenyl)-4-(3-thienyl)-1 H-pyrrolo[2,3-b]pyridine
CAS 1613710-01-2
Quantity 50 mg
Target SNF1LK2/SIK2
Molecular Formula C19H16N2O2S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.