Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARNT/HIF-1 beta Antibody (775146) [HRP], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5630H
Description
ARNT/HIF-1 beta Monoclonal antibody specifically detects ARNT/HIF-1 beta in Human samples. It is validated for Western Blot,ImmunocytochemistrySpecifications
ARNT/HIF-1 beta | |
Monoclonal | |
HRP | |
ARNT, ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, Class E Basic Helix-Loop-Helix Protein 2, Dioxin Receptor, Nuclear Translocator, HIF-1 Beta, HIF1B, HIF-1beta, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, TANGO | |
E. coli-derived recombinant human ARNT/HIF-1β, Asp517-Ala691, Accession # P27540 | |
0.1 mL | |
Angiogenesis, Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
405 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Immunocytochemistry | |
775146 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction