Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARNT/HIF-1 beta Rabbit anti-Human, Clone: 3R0Y4, Novus Biologicals™
SDP

Catalog No. p-7247255 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166610 20 μg
NB166609 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166610 Supplier Novus Biologicals Supplier No. NBP31575720UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ARNT/HIF-1 beta Monoclonal antibody specifically detects ARNT/HIF-1 beta in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ARNT/HIF-1 beta
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3R0Y4
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ARNT, ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, Class E Basic Helix-Loop-Helix Protein 2, Dioxin Receptor, Nuclear Translocator, HIF-1 Beta, HIF1B, HIF-1beta, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, TANGO
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 499-789 of human ARNT/HIF-1 beta (P27540).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 405
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.