Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARNT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109413
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109413 Supplier Thermo Scientific Supplier No. PA5109413
Add to Cart
Add to Cart

ARNT2 Polyclonal Antibody

ARNT2 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen ARNT2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Arnt2
Gene Accession No. Q61324, Q78E60
Gene Alias Ah receptor nuclear translocator; Arnt; ARNT 2; ARNT protein 2; Arnt2; arnt2a; ARNT2b; ARNT2c; arnt2X; Aryl hydrocarbon receptor nuclear translocator 2; aryl hydrocarbon receptor nuclear translocator 2A; aryl hydrocarbon receptor nuclear translocator type 2a; aryl hydrocarbon receptor nuclear translocator type 2b; aryl hydrocarbon receptor nuclear translocator type 2c; aryl-hydrocarbon receptor nuclear translocator 2; bHLHe1; bHLH-PAS factor ARNT2B; chunp6921; Class E basic helix-loop-helix protein 1; Hif-2b; KIAA0307; mKIAA0307; unm t30256; unm_t30256; WEDAS; zfARNT2; zfARNT2a; zfARNT2b; zfARNT2c
Gene Symbols Arnt2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 440-690 of human ARNT2 (NP_055677.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11864, 25243
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.