Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARNT2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986002
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6002 Supplier Bioss Supplier No. BS4238R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AhR, ARNT1, ARNT2 and BMAL1 are members of a family of transcription factors that contain a basic helix-loop-helix motif and a common 'PAS' motif. The aromatic (aryl) hydrocarbon receptor, AhR, is a ligand dependent transcription factor that interacts with specific DNA sequences termed xenobiotic responsive elements (XREs) to activate several genes including CYP1A1, glutathione S-transferase Ya subunit and DT-diaphorase. The Ah receptor nuclear translocator proteins (Arnt 1 or Arnt 2) are required for ligand-dependent nuclear translocation of the Ah receptor and are also necessary for Ah receptor binding to the XRE element. BMAL1 (Brain and Muscle Arnt-like protein 1), also designated Arnt 3, TIC, JAP3 or MOP-3, has been shown to dimerize with Clock and bind to the promoter region of mPer1, suggesting that this protein plays a role in regulation of circadian oscillation in mammals.

Specifications

Antigen ARNT2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Arnt2
Gene Accession No. Q9HBZ2
Gene Alias Ah receptor nuclear translocator; Arnt; ARNT 2; ARNT protein 2; Arnt2; arnt2a; ARNT2b; ARNT2c; arnt2X; Aryl hydrocarbon receptor nuclear translocator 2; aryl hydrocarbon receptor nuclear translocator 2A; aryl hydrocarbon receptor nuclear translocator type 2a; aryl hydrocarbon receptor nuclear translocator type 2b; aryl hydrocarbon receptor nuclear translocator type 2c; aryl-hydrocarbon receptor nuclear translocator 2; BHLHE1; bHLH-PAS factor ARNT2B; chunp6921; Class E basic helix-loop-helix protein 1; Hif-2b; KIAA0307; mKIAA0307; unm t30256; unm_t30256; WEDAS; zfARNT2; zfARNT2a; zfARNT2b; zfARNT2c
Gene Symbols Arnt2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ARNT2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9915
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.