Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aromatase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 8 publications
$207.00 - $501.50
Specifications
Antigen | Aromatase |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 ug/ml, Simple Western reported in scientific literature (PMID 29898754), Immunohistochemistry 1:200. Use reported in scientific literature (PMID 28133606), Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1001596SS
![]() |
Novus Biologicals
NB1001596SS |
0.025 mL |
Each for $207.00
|
|
|||||
NB1001596
![]() |
Novus Biologicals
NB1001596 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
Aromatase Polyclonal specifically detects Aromatase in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Aromatase | |
Western Blot 1 ug/ml, Simple Western reported in scientific literature (PMID 29898754), Immunohistochemistry 1:200. Use reported in scientific literature (PMID 28133606), Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200 | |
Polyclonal | |
Rabbit | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Lipid and Metabolism, Neuroscience | |
PBS with 0.02% Sodium Azide | |
ARO1EC 1.14.14.1, Aromatase, AROsubfamily XIX (aromatization of androgens), CYARMGC104309, CYPXIX, cytochrome P450, family 19, subfamily A, polypeptide 1, Cytochrome P-450AROM, Estrogen synthase, microsomal monooxygenase | |
CYP19A1 | |
IgG | |
Affinity Purified | |
Human and Primate. Immunogen has 91% homology to horse, 90% homology to cow and rabbit, 83% homology to pig, and 81% homology to sheep, goat, and dog proteins. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Primate, Rabbit | |
P11511 | |
1588 | |
A synthetic peptide made to a C-terminal portion of the human Aromatase protein (between residues 400-502). [UniProt# P11511] | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
55 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title