Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aromatase Antibody (H4) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Aromatase |
---|---|
Clone | H4 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:50-1:5000, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Description
Aromatase Monoclonal antibody specifically detects Aromatase in Human, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
Aromatase | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human, Rat, Pig | |
ARO1EC 1.14.14.1, Aromatase, AROsubfamily XIX (aromatization of androgens), CYARMGC104309, CYPXIX, cytochrome P450, family 19, subfamily A, polypeptide 1, Cytochrome P-450AROM, Estrogen synthase, microsomal monooxygenase | |
CYP19A1 | |
IgG2a | |
Protein A purified |
H4 | |
Western Blot 1:50-1:5000, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Lipid and Metabolism, Neuroscience | |
PBS with 0.02% Sodium Azide | |
1588 | |
Synthetic peptide corresponding to AAs 376-390 of human aromatase (KALEDDVIGYPVKK) | |
Primary | |
55 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title