Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aromatase Antibody (H4), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261939PCP
Description
Aromatase Monoclonal antibody specifically detects Aromatase in Human, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Aromatase | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat, Pig | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| H4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| ARO1EC 1.14.14.1, Aromatase, AROsubfamily XIX (aromatization of androgens), CYARMGC104309, CYPXIX, cytochrome P450, family 19, subfamily A, polypeptide 1, Cytochrome P-450AROM, Estrogen synthase, microsomal monooxygenase | |
| Synthetic peptide corresponding to AAs 376-390 of human aromatase (KALEDDVIGYPVKK) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Lipid and Metabolism, Neuroscience | |
| 1588 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction