Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aromatase Recombinant Rabbit Monoclonal Antibody (1H1)
GREENER_CHOICE

Catalog No. PIMA533132
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533132 Supplier Invitrogen™ Supplier No. MA533132
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Aromatase Recombinant Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and catalyzes the last steps of estrogen biosynthesis, three successive hydroxylations of the A ring of androgens. Mutations in this gene can result in either increased or decreased aromatase activity; the associated phenotypes suggest that estrogen functions both as a sex steroid hormone and in growth or differentiation. The gene expresses two transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aromatase
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 1H1
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cyp19a1
Gene Accession No. P11511
Gene Alias Ar; ArKO; ARO; ARO1; Arom; Aromatase; aromatase 1; aromatase cytochrome P450; aromatase cytochrome P450 19A1; CPV1; CYAR; Cyp19; Cyp19a; Cyp19a1; CYP19P1; CYPXIX; CYPXIXA1; Cytochrome P450 17-alpha; cytochrome P450 19 type I; Cytochrome P450 19A1; cytochrome P450 aromatase; cytochrome P-450 aromatase; cytochrome P-450 aromatase (alt.); cytochrome P450 family 19 subfamily a; cytochrome P450 family 19 subfamily A member 1; Cytochrome P450, 19, aromatase; cytochrome P450, family 17, subfamily A, polypeptide 1; cytochrome P450, family 19, subfamily A, polypeptide 1; cytochrome P450, family XIX (conversion of androgen to estrogen), aromatase; cytochrome P450, family19, subfamily A, polypeptide 1; cytochrome P450, subfamily 19; cytochrome P450, subfamily XIX (aromatization of androgens); cytochrome P-450AROM; estrogen synthase; estrogen synthetase; flavoprotein-linked monooxygenase; Int5; Int-5; MGC104309; microsomal monooxygenase; P450AROM; P-450AROM; type III cytochrome p450 aromatase
Gene Symbols Cyp19a1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYP19A1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1588
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.