Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AROS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523220
Description
AROS Polyclonal Antibody for Western Blot
AROS, a nucleolar protein with an unknown function, interacts with extra ribosomal protein RPS19, playing a role in the signaling pathways that regulate rRNA transcription. The deduced amino acid sequence of AROS, derived from cDNA, defines an isoelectric point of 11.3. No known functional motifs were found in AROS except a short polylysine tract embedded in a putative nucleolar localization signal. At a basal level the protein is expressed ubiquitously with a significantly high level of expression in some tissues. Human AROS gene is mapped to chromosome 22q13.1.
Specifications
AROS | |
Polyclonal | |
Unconjugated | |
RPS19BP1 | |
2510038A11Rik; 40S ribosomal protein S19-binding protein 1; Active regulator of SIRT1; AI604923; AROS; dJ1104E15.4; homolog of mouse S19 binding protein; RGD1562823; ribosomal protein S19 binding protein 1; RP5-1104E15.4; RPS19-binding protein 1; RPS19BP1; S19 binding protein; S19BP | |
Rabbit | |
Protein A | |
RUO | |
91582 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q86WX3 | |
RPS19BP1 | |
A portion of amino acids 1-50 of human AROS. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction