Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AROS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560153
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560153 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560153 Supplier Invitrogen™ Supplier No. PA560153
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CRDHLRVNLK FLTRTRSTVA ESVSQQILRQ NRGRKACDRP VAKTKKKKAE GTVFTEEDFQ KFQQEYFGS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.

AROS, a nucleolar protein with an unknown function, interacts with extra ribosomal protein RPS19, playing a role in the signaling pathways that regulate rRNA transcription. The deduced amino acid sequence of AROS, derived from cDNA, defines an isoelectric point of 11.3. No known functional motifs were found in AROS except a short polylysine tract embedded in a putative nucleolar localization signal. At a basal level the protein is expressed ubiquitously with a significantly high level of expression in some tissues. Human AROS gene is mapped to chromosome 22q13.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AROS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPS19BP1
Gene Accession No. Q86WX3
Gene Alias 2510038A11Rik; 40S ribosomal protein S19-binding protein 1; Active regulator of SIRT1; AI604923; AROS; dJ1104E15.4; homolog of mouse S19 binding protein; RGD1562823; ribosomal protein S19 binding protein 1; RP5-1104E15.4; RPS19-binding protein 1; RPS19BP1; S19 binding protein; S19BP
Gene Symbols RPS19BP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AROS. Recombinant protein control fragment (Product #RP-92363).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91582
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.