Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARP10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications
$682.00
Specifications
Antigen | ARP10 |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ARP10 Polyclonal specifically detects ARP10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ARP10 | |
Polyclonal | |
Rabbit | |
Human | |
Q6NTF7 | |
164668 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:CSSCAWELVDFIKAHDHLNLGIFASRLYYHWCKPQQKGLRLLCGSQVPVEVMGFPEFADCWENFVDHEKPLSFNPYKMLEELDKNSRAIKRRLERIKQS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
24 kDa |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
APOBEC-related protein 10, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3H, ARP-10, ARP10Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3H, DNA dC->dU-editing enzyme APOBEC-3H, EC 3.5.4.- | |
APOBEC3H | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title