Learn More
Invitrogen™ Arp2 Recombinant Rabbit Monoclonal Antibody (JE40-62)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550829
Description
Positive control: Jurkat cell lysate, MCF7 cell lysate, SH-SY5Y cell lysate, HeLa cell lysate, human heart tissue, SH-SY5Y, rat spleen tissue lysate, rat liver tissue lysate, rat kidney tissue lysate, RAW264.7 cell lysate, NIH/3T3 cell lysate, C2C12 cell lysate. Predicted band size: 45 kDa Subcellular Location: Cytoplasm, cytoskeleton, Cell projection, Nucleus.
ARP2 (actin-related protein 2) is an ATP-binding component of the ARP2/3 complex - a multiprotein complex that mediates actin polymerization upon stimulation by the nucleation-promoting factor (NPF). The ARP2/3 complex mediates the formation of branched actin networks in the cytoplasm and provides the force for cell motility. The ARP2/3 complex also promotes actin polymerization in the nucleus, thereby regulating gene transcription and repair of damaged DNA.
Specifications
Arp2 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P61160, P61161, Q5M7U6 | |
ACTR2 | |
Synthetic peptide within Human Arp2 aa 345-394/394. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
JE40-62 | |
Unconjugated | |
ACTR2 | |
4921510D23Rik; AA409782; Actin-like protein 14D; Actin-like protein 2; Actin-related protein 14 Da; Actin-related protein 14D; Actin-related protein 2; Actr14D; Actr2; Arp14D; ARP2; ARP2 actin related protein 2 homolog; ARP2 actin-related protein 2; ARP2 actin-related protein 2 homolog; ARP2 actin-related protein 2 homolog (yeast); Arp2-PA; Arp2-PB; Arp2-PC; CG9901; CG9901-PA; CG9901-PB; CG9901-PC; D6Ertd746e; Dmel\CG9901; Dmel_CG9901; I79_003843; LD18955p | |
Rabbit | |
Protein A | |
RUO | |
10097, 289820, 66713 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.