Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arp3 Recombinant Rabbit Monoclonal Antibody (8K4F2)

Arp3 Recombinant Rabbit Monoclonal Antibody (ARC1067)

Supplier:  Invitrogen™ MA535524

Catalog No. PIMA535524


Only null left
Add to Cart

Description

Description

Arp3 Recombinant Monoclonal Antibody for Western Blot

The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Arp3
Recombinant Monoclonal
0.58 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P61158, Q4V7C7, Q99JY9
ACTR3
A synthetic peptide corresponding to a sequence within amino acids 319-418 of human Arp3 (P61158).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
8K4F2
Unconjugated
ACTR3
1200003A09Rik; Actin-like protein 3; actin-related protein 3; actin-related protein 3 homolog (yest); ACTR3; Arp3; ARP3 actin related protein 3 homolog; ARP3 actin-related protein 3; ARP3 actin-related protein 3 homolog; ARP3 actin-related protein 3 homolog (yeast)
Rabbit
Affinity Chromatography
RUO
10096, 74117, 81732
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.