Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARP4 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS5027R

Description
Angiopoietin-like protein 4 (ANGPTL4), is a secreted protein selectively expressed in adipose tissue, liver, and placenta1-3 that plays a variety of roles in vivo, ranging from adipogenesis to angiogenesis to carcinogenesis. The expression of ANGPTL4 is also under nutritional and hormonal control. ANGPTL4 has been observed to induce a strong pro-angiogenic response independent of vascular endothelial growth factor (VEGF), and its expression has been described in hypoxic human tissues as well as a variety of cancers, including liposarcoma, hepatocellular carcinoma, and conventional renal cell carcinoma.Specifications
ARP4 | |
Polyclonal | |
Unconjugated | |
ANGPTL4 | |
425O18-1; angiopoietin like 4; angiopoietin-like 4; Angiopoietin-like protein 4; Angiopoietin-related protein 4; ANGPTL2; Angptl4; ANGPTL4 C-terminal chain; ANGPTL4 N-terminal chain; ARP4; Bk89; Farp; Fasting-i; Fasting-induced adipose factor; Fiaf; fibrinogen/angiopoietin-related protein; HARP; hepatic angiopoietin-related protein; Hepatic fibrinogen/angiopoietin-related protein; HFARP; major histocompatibility complex region NG27; Ng27; NL2; peroxisome proliferator-activated receptor (PPAR) gamma induced angiopoietin-related protein; PGAR; Pgarg; PP1158; PPARG angiopoietin related protein; PSEC0166; Secreted protein Bk89; TGQTL; UNQ171; UNQ171/PRO197 | |
Rabbit | |
Protein A | |
RUO | |
51129 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9BY76 | |
ANGPTL4 | |
KLH conjugated synthetic peptide derived from mouse ARP4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction