Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARPC1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534870
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534870 Supplier Invitrogen™ Supplier No. PA534870
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: ARPC1A transfected 293T lysate. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (89%), Xenopus laevis (91%), Pig (99%), Chicken (96%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ARPC1A is one of seven subunits of the human Arp2/3 protein complex. This subunit is a member of the SOP2 family of proteins and is most similar to the protein ARPC1B. The similarity between these two proteins suggests that they both may function as p41 subunit of the human Arp2/3 complex that has been implicated in the control of actin polymerization in cells. It is possible that the p41 subunit is involved in assembling and maintaining the structure of the Arp2/3 complex. Multiple versions of the p41 subunit may adapt the functions of the complex to different cell types or developmental stages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARPC1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene ARPC1A
Gene Accession No. Q92747
Gene Alias 0610010H08Rik; 1110030K07Rik; 41kDa; AA407347; actin binding protein (Schizosaccharomyces pombe sop2-like); actin related protein 2/3 complex subunit 1A; actin related protein 2/3 complex subunit 1A, 41kDa; actin related protein 2/3 complex, subunit 1A; actin related protein 2/3 complex, subunit 1A, 41kDa; actin-related protein 2/3 complex subunit 1A; ARC1A; Arc40; Arpc1a; epididymis luminal protein 68; epididymis secretory protein Li 307; HEL-68; HEL-S-307; Sid 329; Sid32; Sid329; SOP2Hs; SOP2L; SOP2-like protein; suppressor of profilin/p41 of actin-related complex 2/3
Gene Symbols ARPC1A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ARPC1A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10552
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.