Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARPC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104339
Description
Antibody detects endogenous levels of total ARPC2.
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p34 subunit, has yet to be determined. Two alternatively spliced variants have been characterized to date. Additional alternatively spliced variants have been described but their full length nature has not been determined.
Specifications
ARPC2 | |
Polyclonal | |
Unconjugated | |
ARPC2 | |
2210023N03Rik; 34kDa; actin related protein 2/3 complex subunit 2; actin related protein 2/3 complex subunit 2, 34kDa; actin related protein 2/3 complex, subunit 2; actin related protein 2/3 complex, subunit 2, 34kDa; actin-related protein 2/3 complex subunit 2; actin-related protein 2/3 complex subunit 2 {ECO:0000250; ARC34; Arp2/3 complex 34 kDa subunit; arp2/3 complex 34 kDa subunit {ECO:0000250; ARP2/3 protein complex subunit 34; ARPC2; arpc2 {ECO:0000250; p34-ARC; p34-ARC {ECO:0000250; PNAS-139; PRO2446; testis tissue sperm-binding protein Li 53e; UniProtKB:O15144} | |
Rabbit | |
Affinity chromatography | |
RUO | |
10109, 301511, 76709 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O15144, P85970, Q9CVB6 | |
ARPC2 | |
A synthesized peptide derived from human ARPC2(Accession O15144), corresponding to amino acid residues Q183-N233. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction