Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARPC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521405 Supplier Invitrogen™ Supplier No. PA521405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HeLaS3, mouse kidney, rat kidney. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (87%), Xenopus laevis (92%), Pig (99%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p34 subunit, has yet to be determined. Two alternatively spliced variants have been characterized to date. Additional alternatively spliced variants have been described but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARPC2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene ARPC2
Gene Accession No. O15144, P85970, Q9CVB6
Gene Alias 2210023N03Rik; 34kDa; actin related protein 2/3 complex subunit 2; actin related protein 2/3 complex subunit 2, 34kDa; actin related protein 2/3 complex, subunit 2; actin related protein 2/3 complex, subunit 2, 34kDa; actin-related protein 2/3 complex subunit 2; actin-related protein 2/3 complex subunit 2 {ECO:0000250; ARC34; Arp2/3 complex 34 kDa subunit; arp2/3 complex 34 kDa subunit {ECO:0000250; ARP2/3 protein complex subunit 34; ARPC2; arpc2 {ECO:0000250; p34-ARC; p34-ARC {ECO:0000250; PNAS-139; PRO2446; testis tissue sperm-binding protein Li 53e; UniProtKB:O15144}
Gene Symbols ARPC2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ARPC2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10109, 301511, 76709
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.