Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARPC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582347 Supplier Invitrogen™ Supplier No. PA582347
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVVFSTVFKD DDDVVIGKVF MQEFKEGRRA SHTAPQVLFS HREPPLELKD TDAAVGDNIG YITFVLFPRH TNASARDNTI NLIHTFRDYL HYHIKCSKAY IHTRMRAKTS DFLKVLNRAR PDAEKKEMKT ITGKTFSSR Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p34 subunit, has yet to be determined. Two alternatively spliced variants have been characterized to date. Additional alternatively spliced variants have been described but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARPC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARPC2
Gene Accession No. O15144, P85970, Q9CVB6
Gene Alias 2210023N03Rik; 34kDa; actin related protein 2/3 complex subunit 2; actin related protein 2/3 complex subunit 2, 34kDa; actin related protein 2/3 complex, subunit 2; actin related protein 2/3 complex, subunit 2, 34kDa; actin-related protein 2/3 complex subunit 2; actin-related protein 2/3 complex subunit 2 {ECO:0000250; ARC34; Arp2/3 complex 34 kDa subunit; arp2/3 complex 34 kDa subunit {ECO:0000250; ARP2/3 protein complex subunit 34; ARPC2; arpc2 {ECO:0000250; p34-ARC; p34-ARC {ECO:0000250; PNAS-139; PRO2446; testis tissue sperm-binding protein Li 53e; UniProtKB:O15144}
Gene Symbols ARPC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARPC2. Recombinant protein control fragment (Product #RP-89526).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10109, 301511, 76709
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.