Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARPC3 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299559PCP
Description
ARPC3 Polyclonal antibody specifically detects ARPC3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ARPC3 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
actin related protein 2/3 complex, subunit 3, 21kDa, subunit 3 (21 kD) | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ARPC3. | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
10094 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction